Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000986.3 | NP_000977.1 | 3, 4 | 234 | |
GenBank mRNA | AK311992.1 | - | 3, 4 | 238 | |
BC000690.2 | - | 3, 4 | 213 | 3349215 | |
BC070193.1 | - | 3, 4 | 211 | 6733162 | |
BU754101.1 | - | 3, 4 | 242 | ||
CD513036.1 | - | 2, 3 | 220 | ||
CR456729.1 | - | 2, 3 | 191 | ||
EU668341.1 | - | 3, 4 | 213 | ||
M94314.1 | - | 3, 4 | 230 |