Gene Transcripts

Gene Symbol
RPS19
Entrez Gene ID
6223
Gene Name
ribosomal protein S19
Gene Aliases
DBA, DBA1, S19
Chromosome Location
Chr. 19: 41859918 - 41871416 on Build GRCh38
UniGene ID
Hs.438429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001022.3 NP_001013.1 6 798
NM_001321483.1 NP_001308412.1 6 475
NM_001321484.1 NP_001308413.1 6 605
NM_001321485.1 NP_001308414.1 6 811
GenBank mRNA AK311786.1 - 6 461
BC000023.2 - 6 495 3504083
BC007615.2 - 5 426 3354280
BC017386.1 - 6 464
BC018616.1 - 6 798 4867036
BG398397.1 - 6 605
BU603942.1 - 6 477
CB124389.1 - 6 504
CB125125.1 - 6 475
M81757.1 - 6 448

View Full Product Details