Gene Transcripts

Gene Symbol
SEPT5
Entrez Gene ID
5413
Gene Name
septin 5
Gene Aliases
CDCREL, CDCREL-1, CDCREL1, H5, HCDCREL-1, PNUTL1
Chromosome Location
Chr. 22: 19714464 - 19723322 on Build GRCh38
UniGene ID
Hs.728762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009939.2 NP_001009939.1 4 592
NM_002688.5 NP_002679.2 5 387
GenBank mRNA AK056273.1 - 4 569
AK295940.1 - 5 256
BC025261.1 - 5 351 4622405
BI601927.1 - 4 597
CR456545.1 - 5 267
KJ534756.1 - 5 262
KJ534757.1 - 5 262
U74628.1 - 5 349
U85431.1 - 5 327
Y11593.1 - 5 307

Gene Symbol
SEPT5-GP1BB
Entrez Gene ID
100526833
Gene Name
SEPT5-GP1BB readthrough
Gene Aliases
Chromosome Location
Chr. 22: 19717220 - 19724774 on Build GRCh38
UniGene ID
Hs.283743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037611.1 4 1807
NR_037612.1 4 311
GenBank mRNA AB209168.1 - 4 1807
U59632.1 - 4 311

View Full Product Details