Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005626.4 | NP_005617.2 | 2 | 558 | |
XM_011541951.2 | XP_011540253.1 | 2 | 358 | ||
GenBank mRNA | AB209856.1 | - | 5 | 5498 | |
AK223444.1 | - | 2 | 335 | ||
AK291509.1 | - | 2 | 280 | ||
AK293706.1 | - | 2 | 331 | ||
AK310528.1 | - | 5 | 818 | ||
BC002781.2 | - | 2 | 282 | 3619538 | |
BT007415.1 | - | 2 | 185 | ||
L14076.1 | - | 2 | 232 |