Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005633.3 | NP_005624.2 | 3, 4 | 397 | |
XM_005264515.4 | XP_005264572.1 | 3, 4 | 626 | ||
XM_011533062.2 | XP_011531364.1 | 3, 4 | 565 | ||
XM_011533064.2 | XP_011531366.1 | 3, 4 | 305 | ||
GenBank mRNA | AK290228.1 | - | 4, 5 | 443 | |
AK301960.1 | - | 3, 4 | 456 | ||
L13857.1 | - | 3, 4 | 356 |