Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004139.4 | NP_004130.2 | 11, 12 | 1332 | |
GenBank mRNA | AF105067.1 | - | 11, 12 | 1243 | |
AK313625.1 | - | 11, 12 | 1262 | ||
BC022256.1 | - | 11, 12 | 1261 | 4767724 | |
M35533.1 | - | 11, 12 | 1214 |