Gene Transcripts

Gene Symbol
CDK17
Entrez Gene ID
5128
Gene Name
cyclin dependent kinase 17
Gene Aliases
PCTAIRE2, PCTK2
Chromosome Location
Chr. 12: 96278261 - 96400588 on Build GRCh38
UniGene ID
Hs.506415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170464.2 NP_001163935.1 2, 3 744
NM_002595.4 NP_002586.2 2, 3 744
XM_017019405.1 XP_016874894.1 2, 3 186
XM_017019406.1 XP_016874895.1 2, 3 201
XM_017019407.1 XP_016874896.1 2, 3 631
GenBank mRNA AK290011.1 - 2, 3 636
AK315214.1 - 2, 3 654
BC033005.1 - 2, 3 233 4824359
X66360.1 - 2, 3 181

View Full Product Details