Gene Transcripts

Gene Symbol
PLN
Entrez Gene ID
5350
Gene Name
phospholamban
Gene Aliases
CMD1P, CMH18, PLB
Chromosome Location
Chr. 6: 118548263 - 118561715 on Build GRCh38
UniGene ID
Hs.170839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002667.4 NP_002658.1 2 1238
GenBank mRNA BC005269.1 - 2 1171
M63603.1 - 2 1192

View Full Product Details