Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr. 13: 75620434 - 75859870 on Build GRCh38
UniGene ID
Hs.207631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005358.5 NP_005349.3 1 1369
GenBank mRNA AF330045.1 - 1 1368

View Full Product Details