Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 1324 | |
NM_001145103.1 | NP_001138575.1 | 9 | 1135 | ||
NM_001145104.1 | NP_001138576.1 | 7 | 768 | ||
NM_005902.3 | NP_005893.1 | 9 | 1559 | ||
XM_011521559.2 | XP_011519861.1 | 8 | 1586 | ||
XM_011521560.1 | XP_011519862.1 | 9 | 1498 | ||
GenBank mRNA | AB209616.1 | - | 9 | 1144 | |
AK026690.1 | - | 2 | 957 | ||
AK290881.1 | - | 10 | 1484 | ||
AK298139.1 | - | 9 | 1135 | ||
AK300614.1 | - | 7 | 765 | ||
AK301348.1 | - | 7 | 652 | ||
AK316017.1 | - | 9 | 1184 | ||
BC050743.1 | - | 9 | 1477 | 6094328 | |
CR749287.1 | - | 1 | 320 | ||
U68019.1 | - | 9 | 1327 | ||
U76622.1 | - | 9 | 1298 |