Gene Transcripts

Gene Symbol
TRIP10
Entrez Gene ID
9322
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
CIP4, HSTP, STOT, STP, TRIP-10
Chromosome Location
Chr. 19: 6739682 - 6751526 on Build GRCh38
UniGene ID
Hs.515094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288962.1 NP_001275891.1 3 268
NM_001288963.1 NP_001275892.1 3 268
NM_004240.3 NP_004231.1 3 268
NR_110231.1 3 268
XM_005259683.2 XP_005259740.1 3 268
XM_006722940.1 XP_006723003.1 3 268
GenBank mRNA AB007845.1 - 3 243
AB072596.1 - 3 243
AF380114.1 - 3 188
AF502289.1 - 3 227
AJ000414.1 - 3 227
AK223109.1 - 3 233
AK313296.1 - 3 250
AL832186.1 - 3 354
AY081141.1 - 3 222
BC013002.2 - 3 213 3532036
BG494851.1 - 272
BT006698.1 - 3 188
BT020167.1 - 3 188
BT020171.1 - 3 188
CB997985.1 - 3 281
CR536513.1 - 3 188
DB198268.1 - 3 270

View Full Product Details