Gene Transcripts

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
Chromosome Location
Chr. 2: 232765802 - 232776565 on Build GRCh38
UniGene ID
Hs.467338
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172417.1 NP_001165888.1 2 189
NM_002242.4 NP_002233.2 2 422
GenBank mRNA AF061118.1 - 2 373
AJ006128.1 - 1 328
AJ007557.1 - 2 401
AK314019.1 - 2 422
AY758241.1 - 1 304
BC037290.2 - 2 422 4817527
DA278958.1 - 2 420
DN993097.1 - 2 186

View Full Product Details