Gene Transcripts

Gene Symbol
MED7
Entrez Gene ID
9443
Gene Name
mediator complex subunit 7
Gene Aliases
ARC34, CRSP33, CRSP9
Chromosome Location
Chr. 5: 157138440 - 157142910 on Build GRCh38
UniGene ID
Hs.279902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100816.1 NP_001094286.1 2 1025
NM_004270.4 NP_004261.1 2 741
GenBank mRNA AF031383.1 - 2 699
AF104251.1 - 1 633
AK308393.1 - 2 705
BC005250.1 - 2 695
BC006968.1 - 2 695
BT007368.1 - 1 633
CR457009.1 - 1 633
CR541696.1 - 1 633

View Full Product Details