Gene Transcripts

Gene Symbol
FABP6
Entrez Gene ID
2172
Gene Name
fatty acid binding protein 6
Gene Aliases
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
Chromosome Location
Chr. 5: 160187367 - 160238722 on Build GRCh38
UniGene ID
Hs.519719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040442.1 NP_001035532.1 6 631
NM_001130958.1 NP_001124430.1 7 711
NM_001445.2 NP_001436.1 4 546
GenBank mRNA BC022489.1 - 5 516
DQ132786.1 - 7 712
X90908.1 - 4 546

View Full Product Details