Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006101.2 | NP_006092.1 | 6 | 740 | |
GenBank mRNA | AF017790.1 | - | 6 | 662 | |
AK289396.1 | - | 6 | 694 | ||
AK313184.1 | - | 6 | 692 | ||
BC005239.1 | - | 6 | 710 | ||
BC010171.2 | - | 6 | 668 | ||
BC035617.1 | - | 6 | 671 | 5164424 |