Gene Transcripts

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
Chromosome Location
Chr. 5: 443184 - 471938 on Build GRCh38
UniGene ID
Hs.481464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007277.4 NP_009208.2 4 1066
XM_011513948.2 XP_011512250.1 4 1101
XM_011513949.2 XP_011512251.1 4 1118
XM_011513950.2 XP_011512252.1 3 996
XM_011513951.2 XP_011512253.1 4 1095
XM_017009000.1 XP_016864489.1 4 1101
XM_017009001.1 XP_016864490.1 4 1101
XM_017009002.1 XP_016864491.1 4 1101
XM_017009003.1 XP_016864492.1 4 1101
XM_017009004.1 XP_016864493.1 5 1179
XM_017009005.1 XP_016864494.1 4 1095
XM_017009006.1 XP_016864495.1 4 1095
GenBank mRNA AF055006.1 - 1 118
AK074086.1 - 3 1057
AK131103.1 - 1 547
AK315749.1 - 1 574
AL832434.1 - 2 722
BC001511.2 - 1 478
BC019304.1 - 2 648
BC064569.1 - 3 1008 5590332
GU727644.1 - 4 1066

View Full Product Details