Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr. 9: 100302075 - 100352977 on Build GRCh38
UniGene ID
Hs.494648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161584.1 NP_001155056.1 3 416
NM_017746.3 NP_060216.2 3 472
XM_011518798.1 XP_011517100.1 3 434
XM_017014848.1 XP_016870337.1 3 434
GenBank mRNA AB060968.1 - 3 388
AK000294.1 - 3 388
AK302619.1 - 3 419
AK303662.1 - 3 385
AK304869.1 - 2 189
AY598337.1 - 2 257
BC030652.2 - 3 308 5268105
BP336313.1 - 3 472

View Full Product Details