Gene Transcripts

Gene Symbol
MOB3B
Entrez Gene ID
79817
Gene Name
MOB kinase activator 3B
Gene Aliases
C9orf35, MOB1D, MOBKL2B
Chromosome Location
Chr. 9: 27325209 - 27529852 on Build GRCh38
UniGene ID
Hs.369022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024761.4 NP_079037.3 4 2880
GenBank mRNA AL832572.2 - 4 2763

View Full Product Details