Gene Transcripts

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
Chromosome Location
Chr. 2: 96837914 - 96844021 on Build GRCh38
UniGene ID
Hs.643430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144994.7 NP_659431.5 9 1548
GenBank mRNA AK092564.1 - 8 1422
AY196212.1 - 9 1532

View Full Product Details