Gene Transcripts

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
Chromosome Location
Chr. 1: 32862268 - 32872492 on Build GRCh38
UniGene ID
Hs.524234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171941.2 NP_001165412.1 5 1408
NM_153756.2 NP_715637.2 6 1110
GenBank mRNA AK092102.1 - 6 1043

View Full Product Details