Gene Transcripts

Gene Symbol
VKORC1
Entrez Gene ID
79001
Gene Name
vitamin K epoxide reductase complex subunit 1
Gene Aliases
EDTP308, MST134, MST576, VKCFD2, VKOR
Chromosome Location
Chr. 16: 31090842 - 31094999 on Build GRCh38
UniGene ID
Hs.324844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001311311.1 NP_001298240.1 4 1017
NM_024006.5 NP_076869.1 3 933
NM_206824.2 NP_996560.1 2 823
GenBank mRNA AF176924.1 - 4 957
AK129513.1 - 2 621
AY466113.1 - 3 875
BC000828.1 - 3 663
BC002911.2 - 3 701 3944530
CD249837.1 - 624

View Full Product Details