Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256126.1 | NP_001243055.1 | 11 | 2075 | |
NM_203463.2 | NP_982288.1 | 10 | 2051 | ||
XM_005246440.4 | XP_005246497.1 | 8 | 3366 | ||
XM_017003749.1 | XP_016859238.1 | 8 | 2226 | ||
GenBank mRNA | BC030800.2 | - | 5 | 1282 |