Gene Transcripts
- Gene Symbol
- CLEC2A
- Entrez Gene ID
- 387836
- Gene Name
- C-type lectin domain family 2 member A
- Gene Aliases
- INPE5792, KACL, PILAR, UNQ5792
- Chromosome Location
- Chr. 12: 9882733 - 9932430 on Build GRCh38
- UniGene ID
- Hs.527665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207375.2 |
NP_997258.1
|
4, 5 |
455 |
|
GenBank mRNA |
AY359126.1 |
- |
4, 5 |
421 |
|
View Full Product Details