Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277898.1 | NP_001264827.1 | 7 | 754 | |
NM_008854.5 | NP_032880.1 | 7 | 797 | ||
GenBank mRNA | AK198263.1 | - | 2 | 93 | |
BC003238.1 | - | 7 | 787 | 3497908 | |
BC054834.1 | - | 7 | 782 | 6439034 | |
BU059097.1 | - | 7 | 747 | ||
CD807077.1 | - | 3 | 201 | ||
M12303.1 | - | 7 | 774 |