Gene Transcripts
- Gene Symbol
- Ring1
- Entrez Gene ID
- 19763
- Gene Name
- ring finger protein 1
- Gene Aliases
- Ring1A
- Chromosome Location
- Chr. 17: 34020792 - 34024680 on Build GRCm38
- UniGene ID
- Mm.20343
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC009070.1 |
- |
7 |
1482 |
|
View Full Product Details