Gene Transcripts

Gene Symbol
Sepw1
Entrez Gene ID
20364
Gene Name
selenoprotein W, muscle 1
Gene Aliases
selW
Chromosome Location
Chr. 7: 15917208 - 15922371 on Build GRCm38
UniGene ID
Mm.42829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009156.2 NP_033182.1 4 233
GenBank mRNA AF015284.1 - 4 188
AF241527.2 - 4 186
AK002870.1 - 4 209
AK028124.1 - 4 203
BC052719.1 - 4 195 6814242
BU604713.1 - 4 266
U67890.1 - 4 185

View Full Product Details