Gene Transcripts

Gene Symbol
Pole2
Entrez Gene ID
18974
Gene Name
polymerase (DNA directed), epsilon 2 (p59 subunit)
Gene Aliases
Chromosome Location
Chr. 12: 69201773 - 69228242 on Build GRCm38
UniGene ID
Mm.9199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011133.2 NP_035263.1 7 561
GenBank mRNA AF036898.1 - 7 534
AK077592.1 - 7 562
AK172024.1 - 7 556
BC052170.1 - 7 550
BC063772.1 - 7 550 30147626

View Full Product Details