Gene Transcripts

Gene Symbol
Pou4f3
Entrez Gene ID
18998
Gene Name
POU domain, class 4, transcription factor 3
Gene Aliases
Brn3.1, Brn3c, ddl, dreidel
Chromosome Location
Chr. 18: 42394597 - 42396093 on Build GRCm38
UniGene ID
Mm.488204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138945.2 NP_620395.2 2 290
GenBank mRNA AK039639.1 - 164
BC145941.1 - 2 290 40131244

View Full Product Details