Gene Transcripts
- Gene Symbol
- RGL2
- Entrez Gene ID
- 5863
- Gene Name
- ral guanine nucleotide dissociation stimulator like 2
- Gene Aliases
- HKE1.5, KE1.5, RAB2L
- Chromosome Location
- Chr. 6: 33291654 - 33299388 on Build GRCh38
- UniGene ID
- Hs.509622
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243738.1 |
NP_001230667.1
|
4, 5 |
832 |
|
NM_004761.4 |
NP_004752.1
|
5, 6 |
1029 |
|
XM_011514781.1 |
XP_011513083.1
|
3, 4 |
301 |
|
XM_011514785.2 |
XP_011513087.1
|
3, 4 |
301 |
|
GenBank mRNA |
AB012295.1 |
- |
4, 5 |
381 |
|
AK294442.1 |
- |
4, 5 |
368 |
|
AL050259.1 |
- |
5, 6 |
602 |
|
BC005037.1 |
- |
2, 3 |
70 |
|
BC032681.1 |
- |
5, 6 |
584 |
5585507 |
BX648015.1 |
- |
4, 5 |
374 |
|
View Full Product Details