Gene Transcripts

Gene Symbol
Gpr68
Entrez Gene ID
238377
Gene Name
G protein-coupled receptor 68
Gene Aliases
BB131428, Ogr1
Chromosome Location
Chr. 12: 100876682 - 100908198 on Build GRCm38
UniGene ID
Mm.32160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177673.1 NP_001171144.1 3 1434
NM_001177674.1 NP_001171145.1 3 1616
NM_175493.4 NP_780702.1 3 1737
GenBank mRNA AK036659.1 - 2 1446
AK087410.1 - 3 1418
AK154649.1 - 2 1409
AK155886.1 - 2 1426
BC118034.1 - 2 1415 40106885
BC118044.1 - 2 1415 40106886

View Full Product Details