Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177699.4 | NP_808367.2 | 10 | 1222 | |
GenBank mRNA | AB041045.1 | - | 10 | 1116 | |
AK030737.1 | - | 10 | 1184 | ||
BC017144.1 | - | 9 | 1233 | ||
BC060654.1 | - | 10 | 1205 | 30354865 |