Gene Transcripts

Gene Symbol
Neu1
Entrez Gene ID
18010
Gene Name
neuraminidase 1
Gene Aliases
AA407268, AA407316, Aglp, Apl, Bat-7, Bat7, G9, Map-2, Neu, Neu-1
Chromosome Location
Chr. 17: 34931253 - 34937297 on Build GRCm38
UniGene ID
Mm.8856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010893.3 NP_035023.3 4 903
GenBank mRNA AK077444.1 - 3 2018
AK145680.1 - 4 797
AK190751.1 - 1 27
AK200961.1 - 1 27
BC004666.1 - 4 777 3498197
U93702.1 - 4 779
Y11412.1 - 4 782

View Full Product Details