Gene Transcripts

Gene Symbol
Nsg1
Entrez Gene ID
18196
Gene Name
neuron specific gene family member 1
Gene Aliases
Neep21, m234, p21
Chromosome Location
Chr. 5: 38137192 - 38159467 on Build GRCm38
UniGene ID
Mm.7414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010942.3 NP_035072.2 5 681
GenBank mRNA AF035683.1 - 5 630
AK076451.1 - 5 1121
AK078054.1 - 5 672
AK144503.1 - 5 656
AK148941.1 - 5 681
AX775765.1 - 5 681
BC008272.1 - 5 667 3603440

View Full Product Details