Gene Transcripts

Gene Symbol
Ovol1
Entrez Gene ID
18426
Gene Name
OVO homolog-like 1 (Drosophila)
Gene Aliases
BB147136, Ovo1, movo1
Chromosome Location
Chr. 19: 5549137 - 5560575 on Build GRCm38
UniGene ID
Mm.280225
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019935.3 NP_064319.1 4 2396
GenBank mRNA AK028577.1 - 4 2397
AK132505.1 - 4 2436

View Full Product Details