Gene Transcripts

Gene Symbol
Frat1
Entrez Gene ID
14296
Gene Name
frequently rearranged in advanced T cell lymphomas
Gene Aliases
AW060382
Chromosome Location
Chr. 19: 41829970 - 41832583 on Build GRCm38
UniGene ID
Mm.4573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008043.3 NP_032069.2 1 2441
GenBank mRNA AK164926.1 - 1 1137
U58974.2 - 1 2291

View Full Product Details