Gene Transcripts

Gene Symbol
Gpc4
Entrez Gene ID
14735
Gene Name
glypican 4
Gene Aliases
9530073D23Rik, AF311610, AI385680, AI661372, K-glypican
Chromosome Location
Chr. X: 52050885 - 52164923 on Build GRCm38
UniGene ID
Mm.1528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008150.2 NP_032176.2 3 842
GenBank mRNA BC006622.1 - 3 842
BQ559580.1 - 1 364
CN710713.1 - 1 320
X83577.1 - 3 1163

View Full Product Details