Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285482.1 | NP_001272411.1 | 3 | 1309 | |
NM_001285483.1 | NP_001272412.1 | 3 | 1131 | ||
NM_001285484.1 | NP_001272413.1 | 2 | 1038 | ||
NM_008309.5 | NP_032335.2 | 2 | 1151 | ||
GenBank mRNA | AK082016.1 | - | 2 | 1155 | |
BC103532.1 | - | 2 | 1162 | 40039729 | |
BC103535.1 | - | 3 | 1125 | 40039732 | |
BC103536.1 | - | 3 | 1297 | 40039733 |