Gene Transcripts

Gene Symbol
Hoxa1
Entrez Gene ID
15394
Gene Name
homeobox A1
Gene Aliases
ERA1, Hox-1.6
Chromosome Location
Chr. 6: 52155367 - 52160410 on Build GRCm38
UniGene ID
Mm.197
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010449.4 NP_034579.3 1 631
GenBank mRNA AK083575.1 - 1 632
BC138097.1 - 1 565 8861116
BC138098.1 - 1 568 8861117
M22115.1 - 1 600

View Full Product Details