Gene Transcripts

Gene Symbol
PRCP
Entrez Gene ID
5547
Gene Name
prolylcarboxypeptidase
Gene Aliases
HUMPCP, PCP
Chromosome Location
Chr. 11: 82822940 - 82901740 on Build GRCh38
UniGene ID
Hs.523936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319214.1 NP_001306143.1 5 657
NM_005040.3 NP_005031.1 6 1019
NM_199418.3 NP_955450.2 7 1082
XM_005274093.1 XP_005274150.1 6 749
GenBank mRNA AB451270.1 - 6 818
AB451397.1 - 6 818
AK091786.1 - 7 909
AK302386.1 - 5 608
AK312919.1 - 6 840
BC001500.2 - 6 837 3510081
L13977.1 - 6 847

View Full Product Details