Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301424.1 | NP_001288353.1 | 10 | 1460 | |
NM_007513.4 | NP_031539.3 | 10 | 1576 | ||
GenBank mRNA | AK139342.1 | - | 10 | 1552 | |
AK147991.1 | - | 10 | 1485 | ||
AK163202.1 | - | 10 | 1460 | ||
BC068234.1 | - | 10 | 1467 | ||
BC145779.1 | - | 9 | 1381 | 40131082 | |
BC145781.1 | - | 9 | 1381 | 40131084 | |
M26687.1 | - | 10 | 1494 |