Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007649.4 | NP_031675.1 | 2 | 195 | |
GenBank mRNA | AB196820.1 | - | 2 | 156 | |
AB196821.1 | - | 2 | 156 | ||
AB196822.1 | - | 2 | 156 | ||
AB196823.1 | - | 2 | 156 | ||
AK007741.1 | - | 2 | 196 | ||
AK084984.1 | - | 2 | 224 | ||
AK088027.1 | - | 2 | 242 | ||
AK146173.1 | - | 2 | 242 | ||
AK151601.1 | - | 2 | 186 | ||
BC060977.1 | - | 2 | 223 | 30296173 | |
X17501.1 | - | 2 | 206 | ||
X53526.1 | - | 2 | 206 |