Gene Transcripts

Gene Symbol
Chgb
Entrez Gene ID
12653
Gene Name
chromogranin B
Gene Aliases
Scg-1
Chromosome Location
Chr. 2: 132781278 - 132795072 on Build GRCm38
UniGene ID
Mm.255241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007694.4 NP_031720.1 4 966
GenBank mRNA AK019893.1 - 4 576
AK077489.1 - 4 574
AK133629.1 - 4 570
AK133739.1 - 4 574
AK133786.1 - 4 576
AK158857.1 - 4 967
BC014736.1 - 4 567 4511248
X51429.1 - 4 562
X53028.1 - 4 572

View Full Product Details