Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005415.4 | NP_005406.3 | 2 | 808 | |
XM_017004768.1 | XP_016860257.1 | 1 | 521 | ||
XM_017004769.1 | XP_016860258.1 | 2 | 808 | ||
XM_017004770.1 | XP_016860259.1 | 2 | 779 | ||
XM_017004771.1 | XP_016860260.1 | 2 | 779 | ||
GenBank mRNA | AK312738.1 | - | 2 | 727 | |
BC019944.1 | - | 2 | 715 | 3918690 | |
L20859.1 | - | 2 | 639 |