Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009757.4 | NP_033887.1 | 1 | 628 | |
GenBank mRNA | AF082348.1 | - | 1 | 511 | |
AJ132406.1 | - | 1 | 268 | ||
AK028735.1 | - | 1 | 314 | ||
AK135757.1 | - | 1 | 535 | ||
AK135882.1 | - | 1 | 631 | ||
AK136127.1 | - | 1 | 22 | ||
AK136129.1 | - | 1 | 22 | ||
AK139773.1 | - | 1 | 629 | ||
AK139847.1 | - | 1 | 630 | ||
AK139936.1 | - | 1 | 584 | ||
BC055363.1 | - | 1 | 625 | 30051425 |