Gene Transcripts

Gene Symbol
MAGEC1
Entrez Gene ID
9947
Gene Name
MAGE family member C1
Gene Aliases
CT7, CT7.1
Chromosome Location
Chr. X: 141903856 - 141909401 on Build GRCh38
UniGene ID
Hs.132194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005462.4 NP_005453.2 4 650
XM_011531418.2 XP_011529720.1 4 586
GenBank mRNA AF056334.1 - 612
BC153867.1 - 442

View Full Product Details