Gene Transcripts

Gene Symbol
Zbtb32
Entrez Gene ID
58206
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
4930524C15Rik, FAXF, FAZF, PLZP, Rog, Tzfp
Chromosome Location
Chr. 7: 30589680 - 30602818 on Build GRCm38
UniGene ID
Mm.116789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021397.2 NP_067372.2 6 1708
GenBank mRNA AF241232.1 - 5 1617
AK015881.1 - 4 791

View Full Product Details