Gene Transcripts

Gene Symbol
Rdh14
Entrez Gene ID
105014
Gene Name
retinol dehydrogenase 14 (all-trans and 9-cis)
Gene Aliases
3110030G19Rik, AW215439, PAN2
Chromosome Location
Chr. 12: 10390690 - 10395562 on Build GRCm38
UniGene ID
Mm.119343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023697.2 NP_076186.1 2 616
GenBank mRNA AF303831.1 - 2 631
AK142290.1 - 2 315
AK189191.1 - 1 135
BC020094.1 - 2 604 4014290
BC092299.1 - 2 612 5060293

View Full Product Details