Gene Transcripts

Gene Symbol
Lzic
Entrez Gene ID
69151
Gene Name
leucine zipper and CTNNBIP1 domain containing
Gene Aliases
1810030J04Rik, AW047580
Chromosome Location
Chr. 4: 149485210 - 149496668 on Build GRCm38
UniGene ID
Mm.464434
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026963.5 NP_081239.1 3 854
GenBank mRNA AK007657.1 - 3 343
AK146824.1 - 3 921
AK210899.1 - 3 184
BC023174.1 - 3 854 4952948
DQ058008.1 - 2 179


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK161071.1 - 436 Chr. - - [-]

View Full Product Details