Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133708.3 | NP_598469.1 | 4 | 317 | |
GenBank mRNA | AK007463.1 | - | 4 | 308 | |
AK144311.1 | - | 4 | 582 | ||
AK150473.1 | - | 5 | 453 | ||
AK189202.1 | - | 4 | 277 | ||
AK189514.1 | - | 4 | 344 | ||
AK198797.1 | - | 4 | 277 | ||
AK206464.1 | - | 3 | 223 | ||
AK210576.1 | - | 4 | 336 | ||
AK215045.1 | - | 4 | 335 | ||
BC008116.1 | - | 4 | 276 | 3498021 | |
BY063474.1 | - | 4 | 324 |