Gene Transcripts
- Gene Symbol
- Snapin
- Entrez Gene ID
- 20615
- Gene Name
- SNAP-associated protein
- Gene Aliases
- 25kDa, AA407989, AV026596, Bloc1s7, Snap25bp, Snapap
- Chromosome Location
- Chr. 3: 90488026 - 90491013 on Build GRCm38
- UniGene ID
- Mm.331182
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133854.3 |
NP_598615.1
|
2, 3 |
227 |
|
GenBank mRNA |
AF086838.1 |
- |
2, 3 |
188 |
|
AK007458.1 |
- |
2, 3 |
208 |
|
AK035147.1 |
- |
2, 3 |
224 |
|
AK048007.1 |
- |
2, 3 |
216 |
|
AK050151.1 |
- |
2, 3 |
228 |
|
AK077840.1 |
- |
2, 3 |
218 |
|
AK080718.1 |
- |
2, 3 |
218 |
|
AK086994.1 |
- |
1, 2 |
159 |
|
AK135006.1 |
- |
2, 3 |
219 |
|
AK152057.1 |
- |
2, 3 |
196 |
|
BC048691.1 |
- |
2, 3 |
199 |
6511395 |
View Full Product Details